Gene Transcripts

Gene Symbol
Golgb1
Entrez Gene ID
224139
Gene Name
golgi autoantigen, golgin subfamily b, macrogolgin 1
Gene Aliases
4930428L02Rik, 628101, 6330407A06Rik, AU042952, C130074L01Rik, F730017E11Rik, Gm6840, mKIAA4151
Chromosome Location
Chr. 16: 36875173 - 36933085 on Build GRCm38
UniGene ID
Mm.244815
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030035.1 NP_084311.1 3 305
GenBank mRNA AK020091.1 - 3 194
AK155415.1 - 4 1568
BC061204.1 - 4 346
BC145221.1 - 3 282 9053772
BC150731.1 - 3 282 9087642

View Full Product Details