Gene Transcripts

Gene Symbol
Myrf
Entrez Gene ID
225908
Gene Name
myelin regulatory factor
Gene Aliases
6030439E18, Gm1804, Gm98, Mrf
Chromosome Location
Chr. 19: 10208271 - 10240781 on Build GRCm38
UniGene ID
Mm.329716
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033481.1 NP_001028653.1 11 1686
GenBank mRNA AK141695.1 - 11 1687
BC157942.1 - 8 1077 9007277

View Full Product Details