Gene Transcripts

Gene Symbol
Zyg11a
Entrez Gene ID
230590
Gene Name
zyg-11 family member A, cell cycle regulator
Gene Aliases
BC022150
Chromosome Location
Chr. 4: 108180245 - 108218160 on Build GRCm38
UniGene ID
Mm.301798
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001167936.1 NP_001161408.1 5 1283
GenBank mRNA BC022150.1 - 3 831

View Full Product Details