Gene Transcripts

Gene Symbol
Ntn5
Entrez Gene ID
243967
Gene Name
netrin 5
Gene Aliases
Gm484
Chromosome Location
Chr. 7: 45684022 - 45694556 on Build GRCm38
UniGene ID
Mm.291240
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033356.3 NP_001028528.2 5 1342
NM_001289692.1 NP_001276621.1 5 1342
NM_001289693.1 NP_001276622.1 6 1135
GenBank mRNA AK042348.1 - 6 1137
AK142898.1 - 5 950
BC150901.1 - 5 935 9087812
BC172055.1 - 5 935

View Full Product Details