Gene Transcripts

Gene Symbol
Cypt2
Entrez Gene ID
245566
Gene Name
cysteine-rich perinuclear theca 2
Gene Aliases
Ckt1, Cypt2-ps
Chromosome Location
Chr. X: 105499772 - 105500639 on Build GRCm38
UniGene ID
Mm.250428
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173436.2 NP_775612.2 1, 2 318
GenBank mRNA AF463499.1 - 1, 2 301
BC060953.1 - 1, 2 318 6774045

View Full Product Details