Gene Transcripts

Gene Symbol
Htra4
Entrez Gene ID
330723
Gene Name
HtrA serine peptidase 4
Gene Aliases
B430206E18Rik
Chromosome Location
Chr. 8: 25024928 - 25038962 on Build GRCm38
UniGene ID
Mm.334452
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081187.3 NP_001074656.1 9 2044
GenBank mRNA AV315397.2 - 1 514

View Full Product Details