Gene Transcripts
- Gene Symbol
- Mnda
- Entrez Gene ID
- 381308
- Gene Name
- myeloid cell nuclear differentiation antigen
- Gene Aliases
- A730048F03, Ifi205b, ifi-205-B
- Chromosome Location
- Chr. 1: 173896341 - 173913046 on Build GRCm38
- UniGene ID
- Mm.490248
- Species
- Mus musculus
- Gene Symbol
- Ifi204
- Entrez Gene ID
- 15951
- Gene Name
- interferon activated gene 204
- Gene Aliases
- Ifi16, p204
- Chromosome Location
- Chr. 1: 173747293 - 173767068 on Build GRCm38
- UniGene ID
- Mm.261270
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC010546.1 |
- |
|
289 |
|
CT010351.1 |
- |
|
203 |
|
M31419.1 |
- |
|
330 |
|
View Full Product Details