Gene Transcripts

Gene Symbol
Vmn2r52
Entrez Gene ID
384534
Gene Name
vomeronasal 2, receptor 52
Gene Aliases
EG384534
Chromosome Location
Chr. 7: 10158652 - 10176286 on Build GRCm38
UniGene ID
Mm.389802
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105191.1 NP_001098661.1 6 1934

View Full Product Details