Gene Transcripts

Gene Symbol
Dnaic2
Entrez Gene ID
432611
Gene Name
dynein, axonemal, intermediate chain 2
Gene Aliases
C030015H18Rik, Dnai2, Gm728
Chromosome Location
Chr. 11: 114722210 - 114757889 on Build GRCm38
UniGene ID
Mm.212764
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001034878.2 NP_001030050.2 13 2185
GenBank mRNA EF565270.1 - 12 1801

View Full Product Details