Gene Transcripts

Gene Symbol
Klhl33
Entrez Gene ID
546611
Gene Name
kelch-like 33
Gene Aliases
EG546611
Chromosome Location
Chr. 14: 50891336 - 50896531 on Build GRCm38
UniGene ID
Mm.390692
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001166651.1 NP_001160123.1 1 377

View Full Product Details