Gene Transcripts

Gene Symbol
Dux
Entrez Gene ID
664783
Gene Name
double homeobox
Gene Aliases
AW822073, Dux4, Duxbl, EG664783
Chromosome Location
Chr. : - on Build
UniGene ID
Mm.482575
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081954.1 NP_001075423.1 243

View Full Product Details