Gene Transcripts

Gene Symbol
Cyp4a31
Entrez Gene ID
666168
Gene Name
cytochrome P450, family 4, subfamily a, polypeptide 31
Gene Aliases
Chromosome Location
Chr. 4: 115563626 - 115579015 on Build GRCm38
UniGene ID
Mm.482086
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001252539.1 NP_001239468.1 1 62
NM_201640.3 NP_964002.2 1 62
GenBank mRNA BC013476.1 - 1 13

View Full Product Details