Gene Transcripts

Gene Symbol
Pla2g2c
Entrez Gene ID
29387
Gene Name
phospholipase A2, group IIC
Gene Aliases
Chromosome Location
Chr. 5: 157163646 - 157187654 on Build Rnor_6.0
UniGene ID
Rn.216343
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019202.2 NP_062075.1 4 498
XM_006239142.2 XP_006239204.1 4 516
XM_006239143.2 XP_006239205.1 3 277
XM_008764212.1 XP_008762434.1 4 591
XM_008764213.1 XP_008762435.1 4 369
XM_008764214.1 XP_008762436.1 4 2040
GenBank mRNA BC097325.1 - 4 498
U07798.1 - 4 825

View Full Product Details