Gene Transcripts

Gene Symbol
Rmdn3
Entrez Gene ID
67809
Gene Name
regulator of microtubule dynamics 3
Gene Aliases
1200015F23Rik, AI131757, Fam82a2, Ptpip51, RMD-3, Rmd3
Chromosome Location
Chr. 2: 119136998 - 119157034 on Build GRCm38
UniGene ID
Mm.475909
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033136.3 NP_001028308.1 13 1551
GenBank mRNA AK004786.1 - 13 1477
AK146299.1 - 13 1470
AK151090.1 - 13 1472
AK153322.1 - 13 1472
AK153369.1 - 13 1471
AK171770.1 - 13 1552
BC055754.1 - 13 1445

View Full Product Details