Gene Transcripts

Gene Symbol
Zswim7
Entrez Gene ID
69747
Gene Name
zinc finger SWIM-type containing 7
Gene Aliases
2410012H22Rik
Chromosome Location
Chr. 11: 62267224 - 62281395 on Build GRCm38
UniGene ID
Mm.141150
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027198.1 NP_081474.1 5 714
GenBank mRNA AA027411.1 - 1 306
AK010472.1 - 5 694

View Full Product Details