Gene Transcripts

Gene Symbol
Cdadc1
Entrez Gene ID
71891
Gene Name
cytidine and dCMP deaminase domain containing 1
Gene Aliases
2310010M10Rik, AI449174, AI463398, BB130816, NYD-SP15
Chromosome Location
Chr. 14: 59559388 - 59597984 on Build GRCm38
UniGene ID
Mm.439850
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001168535.1 NP_001162007.1 4 506
NM_001168536.1 NP_001162008.1 4 506
NM_001168537.1 NP_001162009.1 4 506
NM_001168538.1 NP_001162010.1 4 506
NM_027986.3 NP_082262.1 4 506
GenBank mRNA AK009280.1 - 4 498
AK032600.1 - 4 485
AK032813.1 - 4 492
AK038926.1 - 4 486
AK039148.1 - 4 494
AK042494.1 - 4 495
AK045962.1 - 4 492
AK217714.1 - 4 344
BC006901.1 - 4 468 3600168

View Full Product Details