Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_024248.1 | NP_077210.1 | 4 | 488 | |
GenBank mRNA | AK009937.1 | - | 4 | 450 | |
AK038968.1 | - | 4 | 501 | ||
AK041962.1 | - | 4 | 462 | ||
AK054486.1 | - | 4 | 485 | ||
CX204497.1 | - | 4 | 489 |