Gene Transcripts

Gene Symbol
Glb1l
Entrez Gene ID
74577
Gene Name
galactosidase, beta 1-like
Gene Aliases
4833408P15Rik
Chromosome Location
Chr. 1: 75198235 - 75210816 on Build GRCm38
UniGene ID
Mm.458319
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029010.1 NP_083286.1 7 937
GenBank mRNA AK014667.1 - 5 960
AK029925.1 - 7 937
BC021773.1 - 6 690 4222994

View Full Product Details