Gene Transcripts

Gene Symbol
Etnk1
Entrez Gene ID
75320
Gene Name
ethanolamine kinase 1
Gene Aliases
1110061E11Rik, 4930555L11Rik, AI195356, AI841245, C80956, D6Ertd3e, EKI1
Chromosome Location
Chr. 6: 143166866 - 143208546 on Build GRCm38
UniGene ID
Mm.272548
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029250.2 NP_083526.2 8 6287
GenBank mRNA AK004338.1 - 1 306
AK049691.1 - 1 3977
BC023950.1 - 2 3385
BC034743.1 - 2 3315
BC048829.1 - 2 1731

View Full Product Details