Gene Transcripts
- Gene Symbol
- BUB1
- Entrez Gene ID
- 699
- Gene Name
- BUB1 mitotic checkpoint serine/threonine kinase
- Gene Aliases
- BUB1A, BUB1L, hBUB1
- Chromosome Location
- Chr. 2: 110637698 - 110678107 on Build GRCh38
- UniGene ID
- Hs.469649
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001278616.1 |
NP_001265545.1
|
3, 4 |
482 |
|
NM_001278617.1 |
NP_001265546.1
|
4, 5 |
542 |
|
NM_004336.4 |
NP_004327.1
|
4, 5 |
542 |
|
GenBank mRNA |
AF043294.2 |
- |
4, 5 |
462 |
|
AF046078.1 |
- |
4, 5 |
479 |
|
AF047471.1 |
- |
4, 5 |
465 |
|
AF053305.1 |
- |
4, 5 |
493 |
|
AK023540.1 |
- |
4, 5 |
498 |
|
AK225317.1 |
- |
4, 5 |
498 |
|
AK291221.1 |
- |
4, 5 |
495 |
|
AK302418.1 |
- |
3, 4 |
438 |
|
BC017038.1 |
- |
4, 5 |
463 |
|
BC028201.1 |
- |
4, 5 |
463 |
5205385 |
BP313176.1 |
- |
4, 5 |
522 |
|
View Full Product Details