Gene Transcripts

Gene Symbol
Qser1
Entrez Gene ID
99003
Gene Name
glutamine and serine rich 1
Gene Aliases
4732486I23Rik, AI502974
Chromosome Location
Chr. 2: 104754793 - 104816841 on Build GRCm38
UniGene ID
Mm.274314
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001123327.2 NP_001116799.1 12 8754
GenBank mRNA AK029058.1 - 1 2345
AK137781.1 - 1 2254
BC021511.1 - 1 3156

View Full Product Details