Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001285389.2 | NP_001272318.1 | 4 | 491 | |
NM_001285391.2 | NP_001272320.2 | 4 | 814 | ||
NM_001285392.2 | NP_001272321.1 | 4 | 608 | ||
NM_001285393.2 | NP_001272322.1 | 4 | 545 | ||
NM_004414.6 | NP_004405.3 | 4 | 817 | ||
NM_203417.2 | NP_981962.1 | 4 | 603 | ||
NM_203418.2 | NP_981963.1 | 4 | 782 | ||
XM_017028283.1 | XP_016883772.1 | 5 | 1143 | ||
GenBank mRNA | AF400429.1 | - | 4 | 475 | |
AK055845.1 | - | 3 | 1014 | ||
AK092184.1 | - | 4 | 738 | ||
AK131569.1 | - | 4 | 735 | ||
AK299444.1 | - | 4 | 532 | ||
AL556803.3 | - | 4 | 817 | ||
AY325903.1 | - | 4 | 809 | ||
BC002864.2 | - | 4 | 577 | 3944959 | |
BT007363.1 | - | 4 | 521 | ||
BX419260.2 | - | 4 | 735 | ||
CR456878.1 | - | 3 | 443 | ||
CR542065.1 | - | 4 | 521 | ||
U28833.2 | - | 4 | 491 | ||
U85265.2 | - | 4 | 602 | ||
U85266.2 | - | 4 | 545 | ||
U85267.2 | - | 4 | 664 |