Gene Transcripts
- Gene Symbol
- Basp1
- Entrez Gene ID
- 64160
- Gene Name
- brain abundant, membrane attached signal protein 1
- Gene Aliases
- NAP22
- Chromosome Location
- Chr. 2: 77586424 - 77632626 on Build Rnor_6.0
- UniGene ID
- Rn.37779
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_022300.1 |
NP_071636.1
|
2 |
652 |
|
GenBank mRNA |
D14441.1 |
- |
2 |
652 |
|
- Gene Symbol
- LOC100910172
- Entrez Gene ID
- 100910172
- Gene Name
- brain acid soluble protein 1-like
- Gene Aliases
- Chromosome Location
- Chr. 9: 4190678 - 4191953 on Build Rnor_6.0
- Species
- Rattus norvegicus
View Full Product Details