Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001278938.1 | NP_001265867.1 | 7 | 692 | |
NM_004215.4 | NP_004206.1 | 7 | 979 | ||
NM_198120.2 | NP_936056.1 | 7 | 812 | ||
XM_017013960.1 | XP_016869449.1 | 7 | 1175 | ||
GenBank mRNA | AB007619.1 | - | 7 | 860 | |
AF006265.1 | - | 7 | 810 | ||
AK290651.1 | - | 7 | 801 | ||
AY515724.1 | - | 7 | 770 | ||
AY653072.1 | - | 7 | 712 | ||
BC005249.1 | - | 7 | 890 | 3930583 | |
BC017729.1 | - | 7 | 763 | 3453385 | |
BC022506.1 | - | 7 | 877 | 4815654 | |
CR456984.1 | - | 6 | 577 |