Gene Transcripts
- Gene Symbol
- Akr1a1
- Entrez Gene ID
- 78959
- Gene Name
- aldo-keto reductase family 1, member A1 (aldehyde reductase)
- Gene Aliases
- Akr1a4
- Chromosome Location
- Chr. 5: 135482068 - 135498693 on Build Rnor_6.0
- UniGene ID
- Rn.835
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_031000.3 |
NP_112262.1
|
2, 3 |
145 |
|
GenBank mRNA |
BC059133.1 |
- |
2, 3 |
424 |
|
D10854.1 |
- |
2, 3 |
145 |
|
FQ212523.1 |
- |
2, 3 |
387 |
|
FQ212586.1 |
- |
2, 3 |
385 |
|
FQ212779.1 |
- |
2, 3 |
420 |
|
FQ212919.1 |
- |
1, 2 |
95 |
|
FQ213220.1 |
- |
1, 2 |
92 |
|
FQ213467.1 |
- |
2, 3 |
420 |
|
FQ213483.1 |
- |
2, 3 |
416 |
|
FQ213491.1 |
- |
2, 3 |
418 |
|
FQ213579.1 |
- |
2, 3 |
387 |
|
FQ213776.1 |
- |
2, 3 |
387 |
|
FQ214173.1 |
- |
2, 3 |
387 |
|
FQ214441.1 |
- |
2, 3 |
164 |
|
FQ214642.1 |
- |
|
402 |
|
FQ214700.1 |
- |
2, 3 |
360 |
|
FQ215620.1 |
- |
2, 3 |
212 |
|
FQ215772.1 |
- |
2, 3 |
387 |
|
FQ218383.1 |
- |
2, 3 |
387 |
|
FQ218721.1 |
- |
2, 3 |
387 |
|
FQ218729.1 |
- |
2, 3 |
387 |
|
FQ226221.1 |
- |
2, 3 |
420 |
|
FQ228131.1 |
- |
2, 3 |
420 |
|
FQ228132.1 |
- |
2, 3 |
420 |
|
FQ228842.1 |
- |
2, 3 |
416 |
|
FQ229180.1 |
- |
1, 2 |
59 |
|
FQ233156.1 |
- |
2, 3 |
420 |
|
View Full Product Details