Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005164.3 | NP_005155.1 | 1 | 1070 | |
XM_011538027.2 | XP_011536329.1 | 2 | 1010 | ||
XM_017018992.1 | XP_016874481.1 | 1 | 1070 | ||
XM_017018993.1 | XP_016874482.1 | 1 | 1070 | ||
XM_017018994.1 | XP_016874483.1 | 1 | 1070 | ||
GenBank mRNA | AJ000327.1 | - | 1 | 779 | |
AK314254.1 | - | 1 | 1070 | ||
BC104901.1 | - | 1 | 676 | 8143904 | |
BC104903.1 | - | 1 | 670 | 8143906 |