Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_031976.1 | NP_114182.1 | 3 | 496 | |
XM_006249481.2 | XP_006249543.1 | 4 | 496 | ||
GenBank mRNA | BC062008.1 | - | 3 | 382 | |
U42411.1 | - | 3 | 496 | ||
X95577.1 | - | 4 | 466 |