Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_033351.2 | NP_203502.1 | 4 | 1005 | |
XM_006229043.1 | XP_006229105.1 | 5 | 1114 | ||
XM_008759343.1 | XP_008757565.1 | 5 | 1013 | ||
GenBank mRNA | BC061975.1 | - | 5 | 957 | |
FQ229080.1 | - | 4 | 772 | ||
M35495.1 | - | 4 | 1007 | ||
X14323.1 | - | 4 | 1007 |