Gene Transcripts
- Gene Symbol
- Asip
- Entrez Gene ID
- 24152
- Gene Name
- agouti signaling protein
- Gene Aliases
- Chromosome Location
- Chr. 3: 150492010 - 150579870 on Build Rnor_6.0
- UniGene ID
- Rn.205372
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_052979.1 |
NP_443211.1
|
3, 4 |
315 |
|
GenBank mRNA |
AB045587.1 |
- |
3, 4 |
315 |
|
View Full Product Details