Gene Transcripts

Gene Symbol
Zp1
Entrez Gene ID
85271
Gene Name
zona pellucida glycoprotein 1 (sperm receptor)
Gene Aliases
Chromosome Location
Chr. 1: 226966366 - 226972746 on Build Rnor_6.0
UniGene ID
Rn.10890
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053509.1 NP_445961.1 1, 2 208
GenBank mRNA AB000928.1 - 1, 2 208

View Full Product Details