Gene Transcripts

Gene Symbol
Herpud1
Entrez Gene ID
85430
Gene Name
homocysteine-inducible, endoplasmic reticulum stress-inducible, ubiquitin-like domain member 1
Gene Aliases
Sup
Chromosome Location
Chr. 19: 11046903 - 11057570 on Build Rnor_6.0
UniGene ID
Rn.4028
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053523.1 NP_445975.1 2, 3 327
GenBank mRNA AB033771.1 - 2, 3 327
BC061849.1 - 2, 3 349 5621877

View Full Product Details