Gene Transcripts

Gene Symbol
Laptm5
Entrez Gene ID
89783
Gene Name
lysosomal protein transmembrane 5
Gene Aliases
gcd-10
Chromosome Location
Chr. 5: 149047681 - 149069719 on Build Rnor_6.0
UniGene ID
Rn.24799
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053538.2 NP_445990.2 5 411
GenBank mRNA AB046592.1 - 5 423
BC060517.1 - 5 411 5597893
FQ225663.1 - 5 409

View Full Product Details