Gene Transcripts

Gene Symbol
Map2k6
Entrez Gene ID
114495
Gene Name
mitogen-activated protein kinase kinase 6
Gene Aliases
Mkk6
Chromosome Location
Chr. 10: 98707160 - 98823054 on Build Rnor_6.0
UniGene ID
Rn.17256
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053703.3 NP_446155.1 3 223
GenBank mRNA AF369384.1 - 3 223
BC087004.1 - 3 323 7190965

View Full Product Details