Gene Transcripts

Gene Symbol
Cntn5
Entrez Gene ID
114589
Gene Name
contactin 5
Gene Aliases
Nb-2
Chromosome Location
Chr. 8: 7812371 - 8524643 on Build Rnor_6.0
UniGene ID
Rn.10643
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053746.1 NP_446198.1 3 383
XM_008765902.1 XP_008764124.1 3 448
GenBank mRNA D87212.1 - 3 383

View Full Product Details