Gene Transcripts

Gene Symbol
Ccr5
Entrez Gene ID
117029
Gene Name
chemokine (C-C motif) receptor 5
Gene Aliases
Ckr5, Cmkbr5
Chromosome Location
Chr. 8: 133210484 - 133215599 on Build Rnor_6.0
UniGene ID
Rn.10736
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053960.3 NP_446412.2 2 604
GenBank mRNA BC078756.1 - 1 539
Y12009.1 - 2 614

View Full Product Details