Gene Transcripts

Gene Symbol
Fgf22
Entrez Gene ID
170579
Gene Name
fibroblast growth factor 22
Gene Aliases
Chromosome Location
Chr. 7: 12829785 - 12831675 on Build Rnor_6.0
UniGene ID
Rn.198777
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_130751.1 NP_570107.1 3 341
GenBank mRNA AB078902.1 - 3 341

View Full Product Details