Gene Transcripts

Gene Symbol
Pnpla7
Entrez Gene ID
246246
Gene Name
patatin-like phospholipase domain containing 7
Gene Aliases
Nte-related, Ntel, Ntel1
Chromosome Location
Chr. 3: 2182888 - 2261879 on Build Rnor_6.0
UniGene ID
Rn.162643
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144738.2 NP_653339.2 30 3586
GenBank mRNA AY100477.1 - 12 1388
BC083547.1 - 13 1392 7132749

View Full Product Details