Gene Transcripts

Gene Symbol
Snx27
Entrez Gene ID
260323
Gene Name
sorting nexin family member 27
Gene Aliases
Mrt1
Chromosome Location
Chr. 2: 195738613 - 195821608 on Build Rnor_6.0
UniGene ID
Rn.7581
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001110151.1 NP_001103621.1 2 500
NM_152847.2 NP_690060.2 2 500
GenBank mRNA AB010245.1 - 2 500
AB051816.1 - 2 500

View Full Product Details