Gene Transcripts

Gene Symbol
Gzmf
Entrez Gene ID
266704
Gene Name
granzyme F
Gene Aliases
Gzmg, Rnkp7
Chromosome Location
Chr. 15: 35184359 - 35195725 on Build Rnor_6.0
UniGene ID
Rn.10534
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153466.1 NP_703196.1 4 534
GenBank mRNA U57063.1 - 4 534

Gene Symbol
LOC100910060
Entrez Gene ID
100910060
Gene Name
granzyme G-like
Gene Aliases
Chromosome Location
Chr. 15: 35262905 - 35314559 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008770727.1 XP_008768949.1 3 446

View Full Product Details