Gene Transcripts

Gene Symbol
Pycard
Entrez Gene ID
282817
Gene Name
PYD and CARD domain containing
Gene Aliases
Asc
Chromosome Location
Chr. 1: 199438029 - 199439062 on Build Rnor_6.0
UniGene ID
Rn.7817
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172322.1 NP_758825.1 3 481
GenBank mRNA AB053165.1 - 3 481
FQ226280.1 - 3 639
FQ227544.1 - 3 661
FQ227755.1 - 3 639

View Full Product Details