Gene Transcripts

Gene Symbol
WDR5B
Entrez Gene ID
54554
Gene Name
WD repeat domain 5B
Gene Aliases
Chromosome Location
Chr. 3: 122411853 - 122416035 on Build GRCh38
UniGene ID
Hs.567513
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019069.3 NP_061942.2 1 584
GenBank mRNA AK002149.1 - 1 312
AK315186.1 - 1 202
BC025762.1 - 1 584
BC043494.1 - 1 173 6056052
CR457279.1 - 1 77

View Full Product Details