Gene Transcripts
- Gene Symbol
- Ms4a14
- Entrez Gene ID
- 293743
- Gene Name
- membrane-spanning 4-domains, subfamily A, member 14
- Gene Aliases
- Sp3111
- Chromosome Location
- Chr. 1: 227471441 - 227486953 on Build Rnor_6.0
- UniGene ID
- Rn.92466
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_181082.2 |
NP_851596.1
|
5, 6 |
698 |
|
GenBank mRNA |
AY258290.1 |
- |
5, 6 |
698 |
|
View Full Product Details