Gene Transcripts

Gene Symbol
Dpm2
Entrez Gene ID
29640
Gene Name
dolichyl-phosphate mannosyltransferase polypeptide 2, regulatory subunit
Gene Aliases
Chromosome Location
Chr. 3: 11587941 - 11590528 on Build Rnor_6.0
UniGene ID
Rn.167749
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019252.1 NP_062125.1 2 118
GenBank mRNA AB013359.1 - 2 118

View Full Product Details