Gene Transcripts

Gene Symbol
Il3ra
Entrez Gene ID
246144
Gene Name
interleukin 3 receptor, alpha
Gene Aliases
Cyrl
Chromosome Location
Chr. 12: 18534766 - 18542499 on Build Rnor_6.0
UniGene ID
Rn.144673
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006248989.2 XP_006249051.1 7, 8 3287
GenBank mRNA AF030243.1 - 2, 3 491
FQ214452.1 - 4, 5 865

View Full Product Details