Gene Transcripts

Gene Symbol
Gm2a
Entrez Gene ID
282838
Gene Name
GM2 ganglioside activator
Gene Aliases
Chromosome Location
Chr. 10: 40438394 - 40450927 on Build Rnor_6.0
UniGene ID
Rn.98783
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172335.2 NP_758838.2 2 206
GenBank mRNA AB051391.1 - 2 165
BC072474.1 - 2 206 7100875

View Full Product Details