Gene Transcripts

Gene Symbol
Ap2a1
Entrez Gene ID
11771
Gene Name
adaptor-related protein complex 2, alpha 1 subunit
Gene Aliases
Adtaa
Chromosome Location
Chr. 7: 44900373 - 44929490 on Build GRCm38
UniGene ID
Mm.6877
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001077264.1 NP_001070732.1 2 233
NM_007458.2 NP_031484.1 2 233
GenBank mRNA AK135424.1 - 2 162
AK155515.1 - 2 170
AK208311.1 - 1 20
AK212972.1 - 2 138
BC031433.1 - 2 183 4481643
CA327188.1 - 2 239

View Full Product Details