Gene Transcripts

Gene Symbol
Arf6
Entrez Gene ID
11845
Gene Name
ADP-ribosylation factor 6
Gene Aliases
AI788669, AW496366
Chromosome Location
Chr. 12: 69372150 - 69375980 on Build GRCm38
UniGene ID
Mm.27308
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007481.3 NP_031507.1 2 798
GenBank mRNA AK006366.1 - 1 509
AK156960.1 - 2 783
BC003478.1 - 2 760 2655889
BC083112.1 - 2 798 5354797
D87903.1 - 2 740

View Full Product Details