Gene Transcripts

Gene Symbol
Rhod
Entrez Gene ID
11854
Gene Name
ras homolog gene family, member D
Gene Aliases
AI326383, Arhd, Rho, RhoHP1, RhoM
Chromosome Location
Chr. 19: 4425457 - 4439432 on Build GRCm38
UniGene ID
Mm.27701
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007485.4 NP_031511.1 5 879
GenBank mRNA AK008880.1 - 5 880
BC005652.1 - 4 1246
BC047989.1 - 5 888 6333308

View Full Product Details