Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001081437.1 | NP_001074906.1 | 2 | 952 | |
NM_007992.2 | NP_032018.2 | 2 | 952 | ||
GenBank mRNA | AK161522.1 | - | 2 | 896 | |
AK168688.1 | - | 2 | 890 | ||
X75285.1 | - | 2 | 852 |