Gene Transcripts

Gene Symbol
Gata5
Entrez Gene ID
14464
Gene Name
GATA binding protein 5
Gene Aliases
Chromosome Location
Chr. 2: 180325088 - 180335512 on Build GRCm38
UniGene ID
Mm.388880
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008093.2 NP_032119.2 6 2836
GenBank mRNA AY965069.1 - 1 517
U84725.1 - 6 2827

View Full Product Details